4-Amino-N-benzyl-N-methylbenzenesulfonamide

Supplier: Matrix Scientific

023857-500MG
101816-914EA 221.54 USD
101816-914
4-Amino-N-benzyl-N-methylbenzenesulfonamide
4-Amino-N-benzyl-N-methylbenzenesulfonamide

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₂S
MDL Number: MFCD03306016
CAS Number: 108622-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR