(1-Isopropyl-3-pyrrolidinyl)methanol

Supplier: Matrix Scientific

023854-500MG
101816-908EA 203.08 USD
101816-908
(1-Isopropyl-3-pyrrolidinyl)methanol
(1-Isopropyl-3-pyrrolidinyl)methanol

Hazard: Irritant

Formula: C₈H₁₇NO
MDL Number: MFCD08361791
CAS Number: 2148-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR