5,6-Diphenyl-1,2,4-triazin-3-ylamine

Supplier: Matrix Scientific

023853-500MG
101816-906EA 180 USD
101816-906
5,6-Diphenyl-1,2,4-triazin-3-ylamine
5,6-Diphenyl-1,2,4-triazin-3-ylamine

Hazard: Irritant

Formula: C₁₅H₁₂N₄
MDL Number: MFCD00047461
CAS Number: 4511-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR