2-(4-Morpholinylcarbonyl)benzoic acid

Supplier: Matrix Scientific

023852-500MG
101816-904EA 306.15 USD
101816-904
2-(4-Morpholinylcarbonyl)benzoic acid
2-(4-Morpholinylcarbonyl)benzoic acid

Hazard: Irritant

Formula: C₁₂H₁₃NO₄
MDL Number: MFCD00443953
CAS Number: 73728-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR