4-(2-Ethoxyethoxy)aniline hydrochloride

Supplier: Matrix Scientific

023847-500MG
101816-894EA 306.15 USD
101816-894
4-(2-Ethoxyethoxy)aniline hydrochloride
4-(2-Ethoxyethoxy)aniline hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₆ClNO₂
MDL Number: MFCD08559353
CAS Number: 65999-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR