2-Ethoxyacetohydrazide hydrochloride

Supplier: Matrix Scientific

023845-500MG
101816-890EA 143.75 USD
101816-890
2-Ethoxyacetohydrazide hydrochloride
2-Ethoxyacetohydrazide hydrochloride

Hazard: Irritant

Formula: C₄H₁₁ClN₂O₂
MDL Number: MFCD08559360
CAS Number: 39242-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR