N-(2-Methylbenzyl)-2-butanamine hydrochloride

Supplier: Matrix Scientific

023843-500MG
101816-886EA 306.15 USD
101816-886
N-(2-Methylbenzyl)-2-butanamine hydrochloride
N-(2-Methylbenzyl)-2-butanamine hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₀ClN
MDL Number: MFCD07110300
CAS Number: 893590-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR