N-(4-Methylbenzyl)-2-butanamine hydrochloride

Supplier: Matrix Scientific

023842-500MG
101816-884EA 180 USD
101816-884
N-(4-Methylbenzyl)-2-butanamine hydrochloride
N-(4-Methylbenzyl)-2-butanamine hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₀ClN
MDL Number: MFCD07107484
CAS Number: 1049678-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR