4-Oxo-4-(3-oxo-1-piperazinyl)butanoic acid

Supplier: Matrix Scientific

023841-500MG
101816-882EA 438.46 USD
101816-882
4-Oxo-4-(3-oxo-1-piperazinyl)butanoic acid
4-Oxo-4-(3-oxo-1-piperazinyl)butanoic acid

Hazard: Irritant

Formula: C₈H₁₂N₂O₄
MDL Number: MFCD03916517
CAS Number: 590380-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR