N-Butyl-N-cyclopentylamine hydrochloride

Supplier: Matrix Scientific

023840-500MG
101816-880EA 151.8 USD
101816-880
N-Butyl-N-cyclopentylamine hydrochloride
N-Butyl-N-cyclopentylamine hydrochloride

Hazard: Irritant

Formula: C₉H₂₀ClN
MDL Number: MFCD08559402
CAS Number: 1049750-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR