3-Methyl-N-propylcyclohexanamine hydrochloride

Supplier: Matrix Scientific

023838-500MG
101816-876EA 228.85 USD
101816-876
3-Methyl-N-propylcyclohexanamine hydrochloride
3-Methyl-N-propylcyclohexanamine hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₂ClN
MDL Number: MFCD08559399
CAS Number: 1051919-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR