4-Amino-N-(4-fluorophenyl)benzenesulfonamide

Supplier: Matrix Scientific

023832-500MG
101816-866EA 306.15 USD
101816-866
4-Amino-N-(4-fluorophenyl)benzenesulfonamide
4-Amino-N-(4-fluorophenyl)benzenesulfonamide

Hazard: Irritant

Formula: C₁₂H₁₁FN₂O₂S
MDL Number: MFCD01016124
CAS Number: 1494-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR