3,5-Diphenyl-1H-pyrrole-2-carbohydrazide

Supplier: Matrix Scientific

023831-500MG
101816-864EA 306.15 USD
101816-864
3,5-Diphenyl-1H-pyrrole-2-carbohydrazide
3,5-Diphenyl-1H-pyrrole-2-carbohydrazide

Hazard: Irritant

Formula: C₁₇H₁₅N₃O
MDL Number: MFCD02025092
CAS Number: 314763-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR