4-Amino-N-(tert-butyl)benzamide

Supplier: Matrix Scientific

023826-500MG
101816-854EA 306.15 USD
101816-854
4-Amino-N-(tert-butyl)benzamide
4-Amino-N-(tert-butyl)benzamide

Hazard: Irritant

Formula: C₁₁H₁₆N₂O
MDL Number: MFCD00461428
CAS Number: 93483-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR