(4-Aminophenyl)(1-pyrrolidinyl)methanone

Supplier: Matrix Scientific

Synonyms: 4-(Pyrrolidinocarbonyl)aniline

023821-500MG
101816-844EA 52.31 USD
101816-844
(4-Aminophenyl)(1-pyrrolidinyl)methanone
(4-Aminophenyl)(1-pyrrolidinyl)methanone

Hazard: Irritant

Formula: C₁₁H₁₄N₂O
MW: 190.25 g/mol
MDL Number: MFCD02046794
CAS Number: 56302-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR