(6-Iodo-pyridin-2-yl)carbamic acidtert-butyl ester ≥98%

Supplier: Matrix Scientific

023721-250MG
101816-568EA 204.7 USD
101816-568
(6-Iodo-pyridin-2-yl)carbamic acidtert-butyl ester ≥98%
(6-Iodo-pyridin-2-yl)carbamic acidtert-butyl ester

Hazard: Irritant

Formula: C₁₀H₁₃IN₂O₂
MDL Number: MFCD07779510
CAS Number: 849830-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR