Methyl (2S,4S)-4-(4-bromo-2-methylphenoxy)-2-pyrrolidinecarboxylate
Supplier: Matrix Scientific
023709-500MG
101816-538EA
315.38
USD
101816-538
Methyl (2S,4S)-4-(4-bromo-2-methylphenoxy)-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-(4-bromo-2-methylphenoxy)-2-pyrrolidinecarboxylate
Hazard: Irritant
Formula:
C₁₃H₁₆BrNO₃ |
MDL Number:
MFCD08688190 CAS Number: 1217699-69-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...