Methyl (2S,4S)-4-(2-chloro-5-fluorophenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

023705-500MG
101816-530EA 315.38 USD
101816-530
Methyl (2S,4S)-4-(2-chloro-5-fluorophenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(2-chloro-5-fluorophenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₄Cl₂FNO₃
MDL Number: MFCD18803478
CAS Number: 1354487-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR