Methyl (2S,4S)-4-[4-(trifluoromethoxy)phenoxy]-2-pyrrolidinecarboxylate

Supplier: Matrix Scientific

023703-500MG
101816-526EA 315.38 USD
101816-526
Methyl (2S,4S)-4-[4-(trifluoromethoxy)phenoxy]-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-[4-(trifluoromethoxy)phenoxy]-2-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₄F₃NO₄
MDL Number: MFCD08688408
CAS Number: 774576-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR