Methyl (2S,4S)-4-(2,4-dichloro-3,5-dimethylphenoxy)-2-pyrrolidinecarboxylate

Supplier: Matrix Scientific

023699-500MG
101816-518EA 315.38 USD
101816-518
Methyl (2S,4S)-4-(2,4-dichloro-3,5-dimethylphenoxy)-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-(2,4-dichloro-3,5-dimethylphenoxy)-2-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₇Cl₂NO₃
MDL Number: MFCD08688217
CAS Number: 1217662-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR