Methyl (2S,4S)-4-(4-bromo-2-fluorophenoxy)-2-pyrrolidinecarboxylate

Supplier: Matrix Scientific

023695-500MG
101816-510EA 315.38 USD
101816-510
Methyl (2S,4S)-4-(4-bromo-2-fluorophenoxy)-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-(4-bromo-2-fluorophenoxy)-2-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₁₂H₁₃BrFNO₃
MDL Number: MFCD08688187
CAS Number: 1135226-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR