Methyl (2S,4S)-4-(4-chloro-3-ethylphenoxy)-2-pyrrolidinecarboxylate
Supplier: Matrix Scientific
023694-500MG
101816-508EA
315.38
USD
101816-508
Methyl (2S,4S)-4-(4-chloro-3-ethylphenoxy)-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-(4-chloro-3-ethylphenoxy)-2-pyrrolidinecarboxylate
Hazard: Irritant
Formula:
C₁₄H₁₈ClNO₃ |
MDL Number:
MFCD08688204 CAS Number: 1217705-84-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...