Methyl (2S,4S)-4-(4-isopropyl-3-methylphenoxy)-2-pyrrolidinecarboxylate

Supplier: Matrix Scientific

023693-500MG
101816-506EA 315.38 USD
101816-506
Methyl (2S,4S)-4-(4-isopropyl-3-methylphenoxy)-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-(4-isopropyl-3-methylphenoxy)-2-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₁₆H₂₃NO₃
MDL Number: MFCD08688247
CAS Number: 1217756-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR