Methyl (2S,4S)-4-(2,4-difluorophenoxy)-2-pyrrolidinecarboxylate

Supplier: Matrix Scientific

023691-500MG
101816-502EA 235.75 USD
101816-502
Methyl (2S,4S)-4-(2,4-difluorophenoxy)-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-(2,4-difluorophenoxy)-2-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₁₂H₁₃F₂NO₃
MDL Number: MFCD08688222
CAS Number: 1217655-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR