Methyl (2S,4S)-4-(4-nitrophenoxy)-2-pyrrolidinecarboxylate

Supplier: Matrix Scientific

023689-500MG
101816-498EA 235.75 USD
101816-498
Methyl (2S,4S)-4-(4-nitrophenoxy)-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-(4-nitrophenoxy)-2-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₅
MDL Number: MFCD08688302
CAS Number: 218944-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR