Methyl (2S,4S)-4-(8-quinolinyloxy)-2-pyrrolidinecarboxylate

Supplier: Matrix Scientific

023687-500MG
101816-494EA 315.38 USD
101816-494
Methyl (2S,4S)-4-(8-quinolinyloxy)-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-(8-quinolinyloxy)-2-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₆N₂O₃
MDL Number: MFCD08688395
CAS Number: 1135204-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR