Methyl (2S,4S)-4-(3-pyridinyloxy)-2-pyrrolidinecarboxylate dihydrochloride

Supplier: Matrix Scientific

023686-500MG
101816-492EA 315.38 USD
101816-492
Methyl (2S,4S)-4-(3-pyridinyloxy)-2-pyrrolidinecarboxylate dihydrochloride
Methyl (2S,4S)-4-(3-pyridinyloxy)-2-pyrrolidinecarboxylate dihydrochloride

Hazard: Irritant

Formula: C₁₁H₁₆Cl₂N₂O₃
MDL Number: MFCD11100515
CAS Number: 1217679-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR