Methyl (2S,4S)-4-[(6-bromo-2-naphthyl)oxy]-2-pyrrolidinecarboxylate

Supplier: Matrix Scientific

023685-500MG
101816-490EA 235.75 USD
101816-490
Methyl (2S,4S)-4-[(6-bromo-2-naphthyl)oxy]-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-[(6-bromo-2-naphthyl)oxy]-2-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₁₆H₁₆BrNO₃
MDL Number: MFCD08688191
CAS Number: 1217825-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR