Methyl (2S,4S)-4-[(4-chloro-1-naphthyl)oxy]-2-pyrrolidinecarboxylate

Supplier: Matrix Scientific

023683-500MG
101816-486EA 315.38 USD
101816-486
Methyl (2S,4S)-4-[(4-chloro-1-naphthyl)oxy]-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-[(4-chloro-1-naphthyl)oxy]-2-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₁₆H₁₆ClNO₃
MDL Number: MFCD08688212
CAS Number: 1217784-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR