Methyl (2S,4S)-4-(4-ethylphenoxy)-2-pyrrolidinecarboxylate

Supplier: Matrix Scientific

023680-500MG
101816-480EA 235.75 USD
101816-480
Methyl (2S,4S)-4-(4-ethylphenoxy)-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-(4-ethylphenoxy)-2-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₉NO₃
MDL Number: MFCD08688239
CAS Number: 774221-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR