Methyl (2S,4S)-4-(4-methylphenoxy)-2-pyrrolidinecarboxylate

Supplier: Matrix Scientific

023677-500MG
101816-474EA 315.38 USD
101816-474
Methyl (2S,4S)-4-(4-methylphenoxy)-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-(4-methylphenoxy)-2-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₇NO₃
MDL Number: MFCD08688277
CAS Number: 774221-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR