Methyl (2S,4S)-4-(4-isopropylphenoxy)-2-pyrrolidinecarboxylate

Supplier: Matrix Scientific

023675-500MG
101816-470EA 235.75 USD
101816-470
Methyl (2S,4S)-4-(4-isopropylphenoxy)-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-(4-isopropylphenoxy)-2-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₁₅H₂₁NO₃
MDL Number: MFCD08688251
CAS Number: 774221-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR