Methyl (2S,4S)-4-[4-(1,1,3,3-tetramethylbutyl)phenoxy]-2-pyrrolidinecarboxylate

Supplier: Matrix Scientific

023673-500MG
101816-466EA 315.38 USD
101816-466
Methyl (2S,4S)-4-[4-(1,1,3,3-tetramethylbutyl)phenoxy]-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-[4-(1,1,3,3-tetramethylbutyl)phenoxy]-2-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₂₀H₃₁NO₃
MDL Number: MFCD08688407
CAS Number: 1217646-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR