Methyl (2S,4S)-4-(4-propoxyphenoxy)-2-pyrrolidinecarboxylate

Supplier: Matrix Scientific

023668-500MG
101816-456EA 315.38 USD
101816-456
Methyl (2S,4S)-4-(4-propoxyphenoxy)-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-(4-propoxyphenoxy)-2-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₁₅H₂₁NO₄
MDL Number: MFCD08688382
CAS Number: 1217706-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR