Methyl (2S,4S)-4-[4-(benzyloxy)phenoxy]-2-pyrrolidinecarboxylate

Supplier: Matrix Scientific

023666-500MG
101816-452EA 315.38 USD
101816-452
Methyl (2S,4S)-4-[4-(benzyloxy)phenoxy]-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-[4-(benzyloxy)phenoxy]-2-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₁₉H₂₁NO₄
MDL Number: MFCD08688170
CAS Number: 1217634-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR