Methyl (2S,4S)-4-(4-chloro-3-methylphenoxy)-2-pyrrolidinecarboxylate

Supplier: Matrix Scientific

023661-500MG
101816-442EA 235.75 USD
101816-442
Methyl (2S,4S)-4-(4-chloro-3-methylphenoxy)-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-(4-chloro-3-methylphenoxy)-2-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₆ClNO₃
MDL Number: MFCD08688211
CAS Number: 1217844-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR