Methyl (2S,4S)-4-(2-bromo-4-chlorophenoxy)-2-pyrrolidinecarboxylate

Supplier: Matrix Scientific

023660-500MG
101816-440EA 315.38 USD
101816-440
Methyl (2S,4S)-4-(2-bromo-4-chlorophenoxy)-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-(2-bromo-4-chlorophenoxy)-2-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₁₂H₁₃BrClNO₃
MDL Number: MFCD08688185
CAS Number: 1217842-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR