Methyl (2S,4S)-4-(4-fluorophenoxy)-2-pyrrolidinecarboxylate hydrochloride
Supplier: Matrix Scientific
023658-500MG
101816-436EA
538.46
USD
101816-436
Methyl (2S,4S)-4-(4-fluorophenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(4-fluorophenoxy)-2-pyrrolidinecarboxylate hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₅ClFNO₃ |
MDL Number:
MFCD18073234 CAS Number: 753403-75-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...