Methyl (2S,4S)-4-(3,4-dimethylphenoxy)-2-pyrrolidinecarboxylate

Supplier: Matrix Scientific

023654-500MG
101816-428EA 315.38 USD
101816-428
Methyl (2S,4S)-4-(3,4-dimethylphenoxy)-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-(3,4-dimethylphenoxy)-2-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₉NO₃
MDL Number: MFCD08688231
CAS Number: 1217671-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR