Methyl (2S,4S)-4-(3-isopropylphenoxy)-2-pyrrolidinecarboxylate

Supplier: Matrix Scientific

023652-500MG
101816-424EA 315.38 USD
101816-424
Methyl (2S,4S)-4-(3-isopropylphenoxy)-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-(3-isopropylphenoxy)-2-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₁₅H₂₁NO₃
MDL Number: MFCD08688250
CAS Number: 1217666-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR