Methyl (2S,4S)-4-[3-(tert-butyl)phenoxy]-2-pyrrolidinecarboxylate

Supplier: Matrix Scientific

023651-500MG
101816-422EA 235.75 USD
101816-422
Methyl (2S,4S)-4-[3-(tert-butyl)phenoxy]-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-[3-(tert-butyl)phenoxy]-2-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₁₆H₂₃NO₃
MDL Number: MFCD08688200
CAS Number: 1217822-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR