Methyl (2S,4S)-4-(3-iodophenoxy)-2-pyrrolidinecarboxylate

Supplier: Matrix Scientific

023647-500MG
101816-414EA 235.75 USD
101816-414
Methyl (2S,4S)-4-(3-iodophenoxy)-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-(3-iodophenoxy)-2-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₁₂H₁₄INO₃
MDL Number: MFCD08688245
CAS Number: 1217772-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR