Methyl (2S,4S)-4-(3-fluorophenoxy)-2-pyrrolidinecarboxylate

Supplier: Matrix Scientific

023643-500MG
101816-406EA 315.38 USD
101816-406
Methyl (2S,4S)-4-(3-fluorophenoxy)-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-(3-fluorophenoxy)-2-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₁₂H₁₄FNO₃
MDL Number: MFCD08688242
CAS Number: 1217606-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR