Methyl (2S,4S)-4-(3-bromophenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

023642-500MG
101816-404EA 315.38 USD
101816-404
Methyl (2S,4S)-4-(3-bromophenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(3-bromophenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₅BrClNO₃
MDL Number: MFCD11100514
CAS Number: 1217658-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR