Methyl (2S,4S)-4-(2,3,5-trimethylphenoxy)-2-pyrrolidinecarboxylate

Supplier: Matrix Scientific

023634-500MG
101816-388EA 315.38 USD
101816-388
Methyl (2S,4S)-4-(2,3,5-trimethylphenoxy)-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-(2,3,5-trimethylphenoxy)-2-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₁₅H₂₁NO₃
MDL Number: MFCD08688412
CAS Number: 1217649-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR