Methyl (2S,4S)-4-(2,3-dimethylphenoxy)-2-pyrrolidinecarboxylate

Supplier: Matrix Scientific

023633-500MG
101816-386EA 235.75 USD
101816-386
Methyl (2S,4S)-4-(2,3-dimethylphenoxy)-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-(2,3-dimethylphenoxy)-2-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₉NO₃
MDL Number: MFCD08688228
CAS Number: 1217690-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR