Methyl (2S,4S)-4-(2-methylphenoxy)-2-pyrrolidinecarboxylate

Supplier: Matrix Scientific

023632-500MG
101816-384EA 315.38 USD
101816-384
Methyl (2S,4S)-4-(2-methylphenoxy)-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-(2-methylphenoxy)-2-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₇NO₃
MDL Number: MFCD08688275
CAS Number: 1217637-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR