Methyl (2S,4S)-4-(2-ethoxyphenoxy)-2-pyrrolidinecarboxylate

Supplier: Matrix Scientific

023628-500MG
101816-376EA 315.38 USD
101816-376
Methyl (2S,4S)-4-(2-ethoxyphenoxy)-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-(2-ethoxyphenoxy)-2-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₉NO₄
MDL Number: MFCD08688233
CAS Number: 1217679-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR