Methyl (2S,4S)-4-(2,5-dichlorophenoxy)-2-pyrrolidinecarboxylate

Supplier: Matrix Scientific

023625-500MG
101816-370EA 235.75 USD
101816-370
Methyl (2S,4S)-4-(2,5-dichlorophenoxy)-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-(2,5-dichlorophenoxy)-2-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₁₂H₁₃Cl₂NO₃
MDL Number: MFCD08688220
CAS Number: 1217781-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR