Methyl (2S,4S)-4-(2-fluorophenoxy)-2-pyrrolidinecarboxylate
Supplier: Matrix Scientific
023619-500MG
101816-358EA
315.38
USD
101816-358
Methyl (2S,4S)-4-(2-fluorophenoxy)-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-(2-fluorophenoxy)-2-pyrrolidinecarboxylate
Hazard: Irritant
Formula:
C₁₂H₁₄FNO₃ |
MDL Number:
MFCD08688241 CAS Number: 1217734-27-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...